Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PCNA Protein
SDP

Catalog No. NBC118428 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118428 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118428 Supplier Novus Biologicals™ Supplier No. NBC118428
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-261 of Human PCNA The Recombinant Human PCNA Protein is derived from E. coli. The Recombinant Human PCNA Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_002583
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 7.5), 2 mM EDTA, 20% glycerol
Gene ID (Entrez) 5111
Molecular Weight (g/mol) M.W. (theoretical): 28 kDa
Name PCNA Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen 1-261 aa
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Regulatory Status RUO
Gene Symbol PCNA
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Core ESC Like Genes, Stem Cell Markers
Purity or Quality Grade >95%, by SDS-PAGE
Protein PCNA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.