Learn More
Novus Biologicals™ Human Pervanadate Tissue Lysate

Specifications
Specifications
| Tissue | Human Tissue, Pervanadate-treated |
| For Use With (Application) | SDS-Page, Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | In electrophoresis sample buffer (62.5mM Tris pH 6.8, 2% SDS, 5% glycerol, 0.003% bromophenol blue, 0.9% Beta-mercaptoethanol). |
| Host Species | Human |
| Product Type | Human Pervanadate Tissue Lysate |
| Content And Storage | Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.