Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PIP5K2B His Protein
SDP

Catalog No. NBP223327 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223327 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223327 Supplier Novus Biologicals™ Supplier No. NBP223327
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-416 of Human PIP5K2B The Recombinant Human PIP5K2B Protein is derived from E. coli. The Recombinant Human PIP5K2B Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_003550
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 8396
Molecular Weight (g/mol) 49.8kDa
Name PIP5K2B Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen PIP4K2B, 1-416aa Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMSSNCTS TTAVAVAPLS ASKTKTKKKH FVCQKVKLFR ASEPILSVLM WGVNHTINEL SNVPVPVMLM PDDFKAYSKI KVDNHLFNKE NLPSRFKFKE YCPMVFRNLR ERFGIDDQDY QNSVTRSAPI NSDSQGRCGT RFLTTYDRRF VIKTVSSEDV AEMHNILKKY HQFIVECHGN TLLPQFLGMY RLTVDGVETY MVVTRNVFSH RLTVHRKYDL KGSTVAREAS DKEKAKDLPT FKDNDFLNEG QKLHVGEESK KNFLEKLKRD VEFLAQLKIM DYSLLVGIHD VDRAEQEEME VEERAEDEEC ENDGVGGNLL CSYGTPPDSP GNLLSFPRFF GPGEFDPSVD VYAMKSHESS PKKEVYFMAI IDILTPYDTK KKAAHAAKTV KHGAGAEIST VNPEQYSKRF NEFMSNILT
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.