Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Procalcitonin Protein
SDP

Catalog No. NBP199303 Shop All R&D Systems Products
Change view
Click to view available options
0.01mg; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP199303 0.01mg; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP199303 Supplier Novus Biologicals™ Supplier No. NBP199303

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant bioactive protein containing 116 amino acids for Human Procalcitonin The Recombinant Human Procalcitonin Protein is derived from E. coli. The Recombinant Human Procalcitonin Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Accession Number P01258
Formulation Lyophilized from a 10mM Sodium phosphate buffer, pH 7.5.
Gene ID (Entrez) 796
Name Procalcitonin Protein
Purification Method SDS-PAGE
Storage Requirements −20°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >97%, by SDS-PAGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.