Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Prostate Tissue MicroArray (Cancer)
SDP

Catalog No. NBP230169 Shop All Novus Biologicals Products
Change view
Click to view available options
Human prostate cancer tissue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP230169 Human prostate cancer tissue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP230169 Supplier Novus Biologicals™ Supplier No. NBP230169
Add to Cart
Add to Cart

Applications: Immunohistochemistry-Paraffin, Hematoxylin & Eosin Stain

These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 62 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.

  • Species: Human
  • Cross reactivity: Human reactivity reported in scientific literature (PMID: 24865426)
  • Tissue condition: Normal
  • No. of samples: 49
  • No. of patients: 40
  • Core diameter: 2.0mm
  • Section thickness: 4μm

Applications: 

Immunohistochemistry-Paraffin

Recommended Storage

Store at 4°C. Do not freeze.

Warranty and Services

1 Year

Specifications

Quantity 1 Slide
Regulatory Status RUO
Species Human
Tissue Condition Normal
Type Prostate Cancer
For Use With (Application) Immunohistochemistry-Paraffin, Hematoxylin & Eosin Stain
Cross Reactivity Human reactivity reported in scientific literature (PMID: 24865426)

This product is for research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.