Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Proteasome beta 1 His Protein
SDP

Catalog No. NBP137087 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP137087 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP137087 Supplier Novus Biologicals™ Supplier No. NBP137087
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and amino acids 30-241 of PSMB1. The Recombinant Human Proteasome beta 1 Protein is derived from E. coli. The Recombinant Human Proteasome beta 1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_002784
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol
Gene ID (Entrez) 5689
Molecular Weight (g/mol) 27.7kDa
Name Proteasome beta 1 Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MESIRETEVI DPQDLLEGRY FSGALPDDED VVGPGQESDD FELSGSGDLD DLEDSMIGPE VVHPLVPLDN HIPERAGSGS QVPTEPKKLE ENEVIPKRIS PVEESEDVSN KVSMSSTVQG SNIFERTE
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.