Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PSTPIP1 His Protein
SDP

Catalog No. NBP223369 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223369 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223369 Supplier Novus Biologicals™ Supplier No. NBP223369
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-416 of Human PSTPIP1 The Recombinant Human PSTPIP1 Protein is derived from E. coli. The Recombinant Human PSTPIP1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_003969
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 9051
Molecular Weight (g/mol) 50kDa
Name PSTPIP1 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen PSTPIP1, 1-416aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMMPQLQFkDaFWCRDFT AHTGYEVLLQ RLLDGRKMCK DMEELLRQRA QAEERYGKEL VQIARKAGGQ TEINSLRASF DSLKQQMENV GSSHIQLALT LREELRSLEE FRERQKEQRK KYEAVMDRVQ KSKLSLYKKA MESKKTYEQK CRDADDAEQA FERISANGHQ KQVEKSQNKA RQCKDSATEA ERVYRQSIAQ LEKVRAEWEQ EHRTTCEAFQ LQEFDRLTIL RNALWVHSNQ LSMQCVKDDE LYEEVRLTLE GCSIDADIDS FIQAKSTGTE PPAPVPYQNY YDREVTPLTS SPGIQPSCGM IKRFSGLLHG SPKTTSLAAS AASTETLTPT PERNEGVYTA IAVQEIQGNP ASPAQEYRAL YDYTAQNPDE LDLSAGDILE VILEGEDGWW TVERNGQRGF VPGSYLEKL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.