Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human QKI/Quaking His Protein
SDP

Catalog No. NBP223390 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223390 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223390 Supplier Novus Biologicals™ Supplier No. NBP223390
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-341 of Human QKI/Quaking The Recombinant Human QKI/Quaking Protein is derived from E. coli. The Recombinant Human QKI/Quaking Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_006766
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 50% glycerol, 2mM DTT
Gene ID (Entrez) 9444
Molecular Weight (g/mol) 40.1kDa
Name QKI/Quaking Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen QKI, 1-341aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMVGEMET KEKPKPTPDY LMQLMNDKKL MSSLPNFCGI FNHLERLLDE EISRVRKDMY NDTLNGSTEKRSAELPDAVG PIVQLQEKLY VPVKEYPDFN FVGRILGPRG LTAKQLEAET GCKIMVRGKG SMRDKKKEEQ NRGKPNWEHL NEDLHVLITV EDAQNRAEIK LKRAVEEVKK LLVPAAEGED SLKKMQLMEL AILNGTYRDA NIKSPALAFS LAATAQAAPR IITGPAPVLP PAALRTPTPAGPTIMPLIRQ IQTAVMPNGT PHPTAAIVPP GPEAGLIYTP YEYPYTLAPA TSILEYPIEP SGVLGAVATK VRRHDMRVHP YQRIVTADRA ATGN
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.