Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Rap2B His Protein
SDP

Catalog No. NBP223396 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223396 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223396 Supplier Novus Biologicals™ Supplier No. NBP223396
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-183 of Human Rap2B The Recombinant Human Rap2B Protein is derived from E. coli. The Recombinant Human Rap2B Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_002877
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 5912
Molecular Weight (g/mol) 22.9kDa
Name Rap2B Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen RAP2B, 1-183aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMREYKVV VLGSGGVGKS ALTVQFVTGS FIEKYDPTIE DFYRKEIEVD SSPSVLEILD TAGTEQFASM RDLYIKNGQG FILVYSLVNQ QSFQDIKPMR DQIIRVKRYE RVPMILVGNK VDLEGEREVS YGEGKALAEE WSCPFMETSA KNKASVDELF AEIVRQMNYA AQPNGDEGCC SACVIL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.