Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human RASSF1 His Protein
SDP

Catalog No. NBP223397 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223397 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223397 Supplier Novus Biologicals™ Supplier No. NBP223397
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-340 of Human RASSF1 The Recombinant Human RASSF1 Protein is derived from E. coli. The Recombinant Human RASSF1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_009113
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 11186
Molecular Weight (g/mol) 42.9kDa
Name RASSF1 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen RASSF1A, 1-340aa, Human, His tag, E.coli MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMSGE PELIELRELA PAGRAGKGRT RLERANALRI ARGTACNPTR QLVPGRGHRF QPAGPATHTW CDLCGDFIWG VVRKGLQCAH CKFTCHYRCR ALVCLDCCGP RDLGWEPAVE RDTNVDEPVE WETPDLSQAE IEQKIKEYNA QINSNLFMSL NKDGSYTGFI KVQLKLVRPV SVPSSKKPPS LQDARRGPGR GTSVRRRTSF YLPKDAVKHL HVLSRTRARE VIEALLRKFL VVDDPRKFAL FERAERHGQV YLRKLLDDEQ PLRLRLLAGP SDKALSFVLK ENDSGEVNWD AFSMPELHNF LRILQREEEE HLRQILQKYS YCRQKIQEAL HACPLG
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.