Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human RASSF3 His Protein
SDP

Catalog No. NBP223398 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223398 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223398 Supplier Novus Biologicals™ Supplier No. NBP223398
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-238 of Human RASSF3 The Recombinant Human RASSF3 Protein is derived from E. coli. The Recombinant Human RASSF3 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_835463
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 283349
Molecular Weight (g/mol) 30kDa
Name RASSF3 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen RASSF3, 1-238aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMSSGYSS LEEDAEDFFF TARTSFFRRA PQGKPRSGQQ DVEKEKETHS YLSKEEIKEK VHKYNLAVTD KLKMTLNSNG IYTGFIKVQM ELCKPPQTSP NSGKLSPSSN GCMNTLHISS TNTVGEVIEA LLKKFLVTES PAKFALYKRC HREDQVYACK LSDREHPLYL RLVAGPRTDT LSFVLREHEI GEWEAFSLPE LQNFLRILDK EEDEQLQNLK RRYTAYRQKL EEALREVWKP D
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Cancer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.