Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human RPL22 His Protein
SDP

Catalog No. NBP223419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223419 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223419 Supplier Novus Biologicals™ Supplier No. NBP223419
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-128 of Human RPL22 The Recombinant Human RPL22 Protein is derived from E. coli. The Recombinant Human RPL22 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_000974
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 6146
Molecular Weight (g/mol) 17kDa
Name RPL22 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen RPL22, 1-128aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMAPVKKL VVKGGKKKKQ VLKFTLDCTH PVEDGIMDAA NFEQFLQERI KVNGKAGNLG GGVVTIERSK SKITVTSEVP FSKRYLKYLT KKYLKKNNLR DWLRVVANSK ESYELRYFQI NQDEEEEEDE D
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.