Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human SET His Protein
SDP

Catalog No. NBP223464 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223464 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223464 Supplier Novus Biologicals™ Supplier No. NBP223464
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-290 of Human SET The Recombinant Human SET Protein is derived from E. coli. The Recombinant Human SET Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001116293
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 6418
Molecular Weight (g/mol) 35.9kDa
Name SET Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen SET, 1-290aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMAPKRQS PLPPQKKKPR PPPALGPEET SASAGLPKKG EKEQQEAIEH IDEVQNEIDR LNEQASEEIL KVEQKYNKLR QPFFQKRSEL IAKIPNFWVT TFVNHPQVSA LLGEEDEEAL HYLTRVEVTE FEDIKSGYRI DFYFDENPYF ENKVLSKEFH LNESGDPSSK STEIKWKSGK DLTKRSSQTQ NKASRKRQHE EPESFFTWFT DHSDAGADEL GEVIKDDIWP NPLQYYLVPD MDDEEGEGEE DDDDDEEEEG LEDIDEEGDE DEGEEDEDDD EGEEGEEDEG EDD
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.