Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human SGTA His Protein
SDP

Catalog No. NBP144491 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144491 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144491 Supplier R&D Systems™ Supplier No. NBP144491

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-313 of Human SGTA The Recombinant Human SGTA Protein is derived from E. coli. The Recombinant Human SGTA Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_003012
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT
Gene ID (Entrez) 6449
Molecular Weight (g/mol) 35.1kDa
Name SGTA Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MDNKKRLAYA IIQFLHDQLR HGGLSSDAQE SLEVAIQCLE TAFGVTVEDS DLALPQTLPE IFEAAATGKE MPQDLRSPAR TPPSEEDSAE AERLKTEGNE QMKVENFEAA VHFYGKAIEL NPANAVYFCN RAAAYSKLGN YAGAVQDCER AICIDPAYSK AYGRMGLALS SLNKHVEAVA YYKKALELDP DNETYKSNLK IAELKLREAP SPTGGVGSFD IAGLLNNPGF MSMASNLMNN PQIQQLMSGM ISGGNNPLGT PGTSPSQNDL ASLIQAGQQF AQQMQQQNPE LIEQLRSQIR SRTPSASNDD QQELEHHHHH H
Storage Requirements -80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Product Line Novus
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.