Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human SIVA His Protein
SDP

Catalog No. NBP222808 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222808 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222808 Supplier Novus Biologicals™ Supplier No. NBP222808
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-175 of Human SIVA The Recombinant Human SIVA Protein is derived from E. coli. The Recombinant Human SIVA Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_006418
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 10572
Molecular Weight (g/mol) 21.1kDa
Name SIVA Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen SIVA1, 1-175aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMPKRSCP FADVAPLQLK VRVSQRELSR GVCAERYSQE VFEKTKRLLF LGAQAYLDHV WDEGCAVVHLPESPKPGPTG APRAARGQML IGPDGRLIRS LGQASEADPS GVASIACSSC VRAVDGKAVC GQCERALCGQ CVRTCWGCGS VACTLCGLVD CSDMYEKVLC TSCAMFET
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.