Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human SP17 His Protein
SDP

Catalog No. NBP222818 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222818 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222818 Supplier Novus Biologicals™ Supplier No. NBP222818
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-151 of Human SP17 The Recombinant Human SP17 Protein is derived from E. coli. The Recombinant Human SP17 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_059121
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 53340
Molecular Weight (g/mol) 19.8kDa
Name SP17 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen SPA17, 1-151aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMSIPFSN THYRIPQGFG NLLEGLTREI LREQPDNIPA FAAAYFESLL EKREKTNFDP AEWGSKVEDR FYNNHAFEEQ EPPEKSDPKQ EESQISGKEE ETSVTILDSS EEDKEKEEVA AVKIQAAFRG HIAREEAKKM KTNSLQNEEK EENK
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Cancer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.