Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human alpha-Synuclein Protein
SDP

Catalog No. NBP222845 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222845 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222845 Supplier Novus Biologicals™ Supplier No. NBP222845
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-140 of Human alpha-Synuclein The Recombinant Human alpha-Synuclein Protein is derived from E. coli. The Recombinant Human alpha-Synuclein Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_000336
For Use With (Application) SDS-PAGE
Formulation Lyophilized from a 0.2 um filtered 20mM Tris-HCl buffer (pH 7.5), 100mM NaCl, 1mM MgCl2
Gene ID (Entrez) 6622
Molecular Weight (g/mol) 14.4kDa
Name Synuclein-alpha Protein
Purification Method SDS-PAGE
Quantity 1 mg
Source E. Coli
Immunogen alpha-Synuclein, Human, Recombinant, E.coli - Lyophilized MDVFMKGLSK AKEGVVAAAE KTKQGVAEAA GKTKEGVLYV GSKTKEGVVH GVATVAEKTK EQVTNVGGAV VTGVTAVAQK TVEGAGSIAA ATGFVKKDQL GKNEEGAPQE GILEDMPVDP DNEAYEMPSE EGYQDYEPEA
Storage Requirements 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.