Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human TBC1D1 His Protein
SDP

Catalog No. NBP222851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222851 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222851 Supplier Novus Biologicals™ Supplier No. NBP222851
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-265 of Human TBC1D1 The Recombinant Human TBC1D1 Protein is derived from E. coli. The Recombinant Human TBC1D1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001240842
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 23216
Molecular Weight (g/mol) 33.2kDa
Name TBC1D1 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen TBC1D1, 1-265aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMSEEEAF KMLKFLMFDM GLRKQYRPDM IILQIQMYQL SRLLHDYHRD LYNHLEEHEI GPSLYAAPWF LTMFASQFPL GFVARVFDMI FLQGTEVIFK VALSLLGSHK PLILQHENLE TIVDFIKSTL PNLGLVQMEK TINQVFEMDI AKQLQAYEVE YHVLQEELID SSPLSDNQRM DKLEKTNSSL RKQNLDLLEQ LQVANGRIQS LEATIEKLLS SESKLKQAML TLELERSALL QTVEELRRRS AEPSDREPEC TQPEPTGD
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.