Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human TC21/R-Ras2 His Protein
SDP

Catalog No. NBP130244 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130244 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130244 Supplier Novus Biologicals™ Supplier No. NBP130244
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1- 201 of Human TC21/R-Ras2 The Recombinant Human TC21/R-Ras2 Protein is derived from E. coli. The Recombinant Human TC21/R-Ras2 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_036382
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 1mM DTT, 10% glycerol, 2mM EDTA
Gene ID (Entrez) 22800
Molecular Weight (g/mol) 25.2kDa
Name TC21/R-Ras2 Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MGSSHHHHHH SSGLVPRGSH MAAAGWRDGS GQEKYRLVVV GGGGVGKSAL TIQFIQSYFV TDYDPTIEDS YTKQCVIDDR AARLDILDTA GQEEFGAMRE QYMRTGEGFL LVFSVTDRGS FEEIYKFQRQ ILRVKDRDEF PMILIGNKAD LDHQRQVTQE EGQQLARQLK VTYMEASAKI RMNVDQAFHE LVRVIRKFQE QECPPSPEPT RKEKDKKGCH C
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.