Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Thioredoxin Reductase 1/TRXR1 His Protein
SDP

Catalog No. NBP144456 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144456 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144456 Supplier Novus Biologicals™ Supplier No. NBP144456
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 161-647 of Human Thioredoxin Reductase 1/TRXR1 The Recombinant Human Thioredoxin Reductase 1/TRXR1 Protein is derived from E. coli. The Recombinant Human Thioredoxin Reductase 1/TRXR1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Accession Number Q16881
For Use With (Application) Bioactivity, SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 7296
Molecular Weight (g/mol) 55.7kDa
Name Thioredoxin Reductase 1/TRXR1 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen TXNRD1, 161-647aa, Human, His tag, E.coli (NP_001087240).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.