Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human U2AF35 His Protein
SDP

Catalog No. NBP222915 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222915 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222915 Supplier Novus Biologicals™ Supplier No. NBP222915
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-240 of Human U2AF35 The Recombinant Human U2AF35 Protein is derived from E. coli. The Recombinant Human U2AF35 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001020374
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 199746
Molecular Weight (g/mol) 30.3kDa
Name U2AF1L4 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen U2AF1, 1-240aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMAEYLAS IFGTEKDKVN CSFYFKIGAC RHGDRCSRLH NKPTFSQTIL IQNIYRNPQN SAQTADGSHC AVSDVEMQEH YDEFFEEVFT EMEEKYGEVE EMNVCDNLGD HLVGNVYVKF RREEDAEKAV IDLNNRWFNG QPIHAELSPV TDFREACCRQ YEMGECTRGG FCNFMHLKPI SRELRRELYG RRRKKHRSRS RSRERRSRSR DRGRGGGGGG GGGGGGRERD RRRSRDRERS GRF
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.