Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human UQCRC2 His Protein
SDP

Catalog No. NBP222916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222916 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222916 Supplier Novus Biologicals™ Supplier No. NBP222916
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 15-453 of Human UQCRC2 The Recombinant Human UQCRC2 Protein is derived from E. coli. The Recombinant Human UQCRC2 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_003357
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 2M UREA, 10% glycerol, 0.1 M NaCl
Gene ID (Entrez) 7385
Molecular Weight (g/mol) 49kDa
Name UQCRC2 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen UQCRC2, 15-453aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MSLKVAPKVK ATAAPAGAPP QPQDLEFTKL PNGLVIASLE NYSPVSRIGL FIKAGSRYED FSNLGTTHLL RLTSSLTTKG ASSFKITRGI EAVGGKLSVT ATRENMAYTV ECLRGDVDIL MEFLLNVTTA PEFRRWEVAD LQPQLKIDKA VAFQNPQTHV IENLHAAAYR NALANPLYCP DYRIGKVTSE ELHYFVQNHF TSARMALIGL GVSHPVLKQV AEQFLNMRGG LGLSGAKANY RGGEIREQNG DSLVHAAFVA ESAVAGSAEA NAFSVLQHVL GAGPHVKRGS NTTSHLHQAV AKATQQPFDV SAFNASYSDS GLFGIYTISQ ATAAGDVIKA AYNQVKTIAQ GNLSNTDVQA AKNKLKAGYL MSVESSECFL EEVGSQALVA GSYMPPSTVL QQIDSVANAD IINAAKKFVS GQKSMAASGN LGHTPFVDEL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.