Learn More
Description
Specifications
Specifications
| Accession Number | AAH16057 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0), 20% glycerol |
| Gene ID (Entrez) | 10083 |
| Molecular Weight (g/mol) | 64.6kDa |
| Name | USH1C Protein |
| Purification Method | Protein |
| Immunogen | MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMDR KVAREFRHKV DFLIENDAEK DYLYDVLRMY HQTMDVAVLV GDLKLVINEP SRLPLFDAIR PLIPLKHQVE YDQLTPRRSR KLKEVRLDRL HPEGLGLSVR GGLEFGCGLF ISHLIKGGQA DSVGLQVGDE IVRINGYSIS SCTHEEVINL IRTKKTVSIK VRHIGLIPVK SSPDEPLTWQ YVDQFVSESG GVRGSLGSPG NRENKEKKVF ISLVGSRGLG CSISSGPIQK PGIFISHVKP GSLSAEVGLE IGDQIVEVNG VDFSNLDHKE GRELFMTDRE RLAEARQREL QRQELLMQKR LAMESNKILQ EQQEMERQRR KEIAQKAAEE NERYRKEMEQ IVEEEEKFKK QWEEDWGSKE QLLLPKTITA EVHPVPLRKP KYDQGVEPEL EPADDLDGGT EEQGEQDFRK YEEGFDPYSM FTPEQIMGKD VRLLRIKKEG SLDLALEGGV DSPIGKVVVS AVYERGAAER HGGIVKGDEI MAINGKIVTD YTLAEADAAL QKAWNQGGDW IDLVVAVCPP KEYDDELTFF |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
