Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Wnt-7a His Protein
SDP

Catalog No. NBP222946 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222946 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222946 Supplier Novus Biologicals™ Supplier No. NBP222946
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 32-349 of Human Wnt-7a The Recombinant Human Wnt-7a Protein is derived from E. coli. The Recombinant Human Wnt-7a Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_004616
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 7476
Molecular Weight (g/mol) 38kDa
Name Wnt-7a Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen WNT7A, 32-349aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSLGASIIC NKIPGLAPRQ RAICQSRPDA IIVIGEGSQM GLDECQFQFR NGRWNCSALG ERTVFGKELKVGSREAAFTY AIIAAGVAHA ITAACTQGNL SDCGCDKEKQ GQYHRDEGWK WGGCSADIRY GIGFAKVFVD AREIKQNART LMNLHNNEAG RKILEENMKL ECKCHGVSGS CTTKTCWTTL PQFRELGYVL KDKYNEAVHV EPVRASRNKR PTFLKIKKPL SYRKPMDTDL VYIEKSPNYCEEDPVTGSVG TQGRACNKTA PQASGCDLMC CGRGYNTHQY ARVWQCNCKF HWCCYVKCNT CSERTEMYTC K
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.