Learn More
Novus Biologicals™ Imidazoquinoline Resiquimod (R-848), TLR7 and TLR8 Ligand
Shop All Novus Products

Description
Imidazoquinoline Resiquimod (R-848), TLR7 and TLR8 ligand is a low molecular weight synthetic molecule of the Imidazoquinoline family. It has potent immunostimulatory activity. It stimulates antibody secretion, cytokine production, protection from apoptosis and CD80 up-regulation.
- Species: Human
- Stimulation of TLR8 has been achieved with 0.5-5μg/mL
Recommended Storage
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Specifications
Specifications
| Buffer | DMSO |
| Content And Storage | Store at -20°C. |
| For Use With (Application) | Identity determined by MS, 1H-NMR and 13C-NMR. This product is useful for activation of mouse TLR7 and human TLR7 and TLR8. Stimulation of TLR8 has been achieved with 0.5-5 ug/mL. Use in functional assays reported in scientific literature (PMID 26134251). |
| Product Type | Imidazoquinoline Resiquimod (R-848), TLR7 and TLR8 Ligand |
| Purity | >98%, by HPLC. |
| Quantity | 0.5 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.