Learn More
Specifications
Specifications
| For Use With (Application) | Flow Cytometry, Flow (Intracellular) |
| Product Type | Intracellular Receptor Staining Flow Assay Kit |
| Content And Storage | Fixation buffer 1X: 1 X 60mL, Permeabilization buffer 10X: 2 X 60mL, Staining buffer 1X: 2 X 60mL, Brefeldin A 1000X: 1 X 100μL. Storage is content dependent. |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
