Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 9365 |
| Species | Human |
| Product Line | Novus |
| Purity | >50%, by HPLC |
| Accession Number | NP_004786.2 |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | Lyophilized from a 5 pmol/ul solution in 5% (v/v) acetonitrile/H2O and TFA salt. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-2432. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | EC 3.2.1, EC 3.2.1.31, klotho |
| Gene Symbol | KL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
