Learn More
Novus Biologicals™ Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody Blocking Peptide
Shop All R&D Systems Products

Description
Specifications
Specifications
| Gene ID (Entrez) | 23135 |
| Species | Human |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-06640. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6 B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6 B, Lysine demethylase 6 B |
| Gene Symbol | KDM6B |
| Product Type | Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 |
| Quantity | 0.05 mg |
| Regulatory Status | RUO |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.