Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 4544 |
| Species | Human |
| Accession Number | P49286 |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | PBS, pH 7.4 |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NLS932. To Block The Specific Antibody Activity in Immunohistochemistry Analysis, Pre-Incubate The Antibody with a 10-100 x Molar Excess of Peptide for 60 Minutes (at Room Temperature. Optimization of The Molar Ratio of Antigen to Antibody May be Required. Centrifugation of The Pre-Adsorbed Solution Prior to Addition to The Slide May Decrease Background Due to Non-Specific Binding of Large Antibody-Antigen Complexes. For Further Blocking Peptide Related Protocol, Click Here .) |
| Gene Alias | FGQTL2, Mel 1 b melatonin receptor, Mel1b receptor, MEL-1 B-R, melatonin receptor 1 B, melatonin receptor MEL1B, melatonin receptor type 1 B, MT2 |
| Gene Symbol | MTNR1B |
| Product Type | Melatonin Receptor 1B |
| Quantity | 0.05 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
