Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Methionine Sulfoxide Reductase B Protein
SDP

Catalog No. NBP14930205 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14930205 0.05 mg
NBP14930202 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP14930205 Supplier Novus Biologicals™ Supplier No. NBP1493020.05MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-116 of Human Methionine Sulfoxide Reductase B The Recombinant Human Methionine Sulfoxide Reductase B Protein is derived from E. coli. The Recombinant Human Methionine Sulfoxide Reductase B Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl Buffer (pH 7.5) containing 1mM DTT, 0.1mM PMSF, 2mM EDTA, 10% Glycerol
Gene ID (Entrez) 51734
Molecular Weight (g/mol) 14.8kDa
Purification Method Protein
Quantity 0.05 mg
Source Human
Immunogen SEPX1(U95C), 1-116aa. MGSSHHHHHHSSGLVPRGSHMSFCSFFGGEVFQNHFEPGVYVCAKCGYELFSSRSKYAHSSPWPAFTETIHADSVAKRPEHNRSEALKVSCGKCGNGLGHEFLNDGPKPGQSRFCIFSSSLKFVPKGKETSASQGH
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Methionine Sulfoxide Reductase B
Conjugate Unconjugated
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.