Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Midkine His Protein
SDP

Catalog No. NBP130185 Shop All R&D Systems Products
Change view
Click to view available options
:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP130185 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130185 Supplier Novus Biologicals™ Supplier No. NBP130185
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 21-143 of Human Midkine The Recombinant Human Midkine Protein is derived from E. coli. The Recombinant Human Midkine Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number P21741
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 0.1 M NaCl, 2mM DTT, and 20% Glycerol
Gene ID (Entrez) 4192
Molecular Weight (g/mol) 15.7kDa
Name Midkine Protein
Purification Method Protein
Quantity 0.1mg
Source E. Coli
Immunogen Midkine, 21-143 aa, Human, His-tagged, Recombinant, E.coli (NP_0023820.
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.