Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse AlphaB Crystallin/CRYAB Protein
SDP

Catalog No. NBC118376 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118376 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118376 Supplier Novus Biologicals™ Supplier No. NBC118376
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-175 of Mouse AlphaB Crystallin/CRYAB The Recombinant Mouse AlphaB Crystallin/CRYAB Protein is derived from E. coli. The Recombinant Mouse AlphaB Crystallin/CRYAB Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 10 % glycerol
Gene ID (Entrez) 1410
Molecular Weight (g/mol) M.W. (theoretical): 20 kDa
Name AlphaB Crystallin/CRYAB protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MDIAIHHPWIRRPFFPFHSPSRLFDQFFGEHLLESDLFSTATSLSPFYLRPPSFLRAPSWIDTGLSEMRLEKDRFSVNLDVKHFSPEELKVKVLGDVIEVHGKHEERQDEHGFISREFHRKYRIPADVDPLTITSSLSSDGVLTVNGPRKQVSGPERTIPITREEKPAVAAAPKK
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Gene Symbol CRYAB
Cross Reactivity Mouse
Purity or Quality Grade >95%, by SDS-PAGE
Protein AlphaB Crystallin/CRYAB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.