Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse CCL2/MCP1 His Protein
SDP

Catalog No. NBP222772 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222772 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222772 Supplier Novus Biologicals™ Supplier No. NBP222772
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 24-148 of Mouse His-tag CCL2/MCP1 The Recombinant Mouse CCL2/MCP1 Protein is derived from E. coli. The Recombinant Mouse CCL2/MCP1 Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) In vivo Assay, SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 6347
Molecular Weight (g/mol) 16kDa
Name CCL2/MCP1 protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen CCL2, 24-148aa, Mouse His tag, E.coli MGSSHHHHHH SSGLVPRGSH MQPDAVNAPL TCCYSFTSKM IPMSRLESYK RITSSRCPKE AVVFVTKLKR EVCADPKKEW VQTYIKNLDR NQMRSEPTTL FKTASALRSS APLNVKLTRK SEANASTTFS TTTSSTSVGV TSVTVN
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.