Learn More
Description
Specifications
Specifications
| Accession Number | NP_005893 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH8.0), 20% glycerol, 0.1 M NaCl and 1mM DTT |
| Gene ID (Entrez) | 4088 |
| Molecular Weight (g/mol) | 50.2kDa |
| Name | Smad3 Protein |
| Purification Method | Protein |
| Immunogen | MGSSHHHHHH SSGLVPRGSH MSSILPFTPP IVKRLLGWKK GEQNGQEEKW CEKAVKSLVK KLKKTGQLDE LEKAITTQNV NTKCITIPRS LDGRLQVSHR KGLPHVIYCR LWRWPDLHSH HELRAMELCE FAFNMKKDEV CVNPYHYQRV ETPVLPPVLV PRHTEIPAEF PPLDDYSHSI PENTNFPAGI EPQSNIPETP PPGYLSEDGE TSDHQMNHSM DAGSPNLSPN PMSPAHNNLD LQPVTYCEPA FWCSISYYEL NQRVGETFHA SQPSMTVDGF TDPSNSERFC LGLLSNVNRN AAVELTRRHI GRGVRLYYIG GEVFAECLSD SAIFVQSPNC NQRYGWHPAT VCKIPPGCNL KIFNNQEFAA LLAQSVNQGF EAVYQLTRMC TIRMSFVKGW GAEYRRQTVT STPCWIELHL NGPLQWLDKV LTQMGSPSIR CSSVS |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
