Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mycobacterium Tuberculosis ESAT6 Protein
SDP

Catalog No. NBP19904505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19904505 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP19904505 Supplier Novus Biologicals™ Supplier No. NBP1990450.05MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-95 of Mycobacterium tuberculosis Mycobacterium Tuberculosis ESAT6 The Recombinant Bacteria Mycobacterium Tuberculosis ESAT6 Protein is derived from E. coli. The Recombinant Bacteria Mycobacterium Tuberculosis ESAT6 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_338543.1
Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing, 10% glycerol
Molecular Weight (g/mol) 11kDa
Purification Method SDS-PAGE
Quantity 50 μg
Immunogen ESAT6, 1-95 aa, Mycobacterium tuberculosis, His tag, Baculovirus (NP_338543.1).
Storage Requirements Store at 4°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Mycobacterium Tuberculosis ESAT6
Conjugate Unconjugated
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.