Learn More
Novus Biologicals™ MYH Overexpression Lysate

Description
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC21376 and based on accession number NM_148174. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens mutY homolog (E. coli) (MUTYH), transcript variant beta3, mRNA. |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Research Category | Cancer |
| Target | MYH |
| Synonym | CYP2C, EC 3.2.2, EC 3.2.2.-, hMYH, MGC4416, mutY (E. coli) homolog, MutY homolog, mutY homolog (E. coli), Muyth, MYHA/G-specific adenine DNA glycosylase |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.