Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 6866 |
| Species | Mouse |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB300-201. Blocking/Neutralizing Was Reported in Scientific Literature (PMID: 20038444). IHC-Fr Was Reported in Scientific Literature (PMID 24735328) For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | gamma tachykinin 3, neurokinin beta, neurokinin B-like, neuromedin K, preprotachykinin B, PRO1155, tachykinin 3, tachykinin-3, ZNEUROK1NKNBNKB |
| Gene Symbol | TAC3 |
| Product Type | Neurokinin B |
| Quantity | 0.05 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
