Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Normal Goat Serum
SDP

Catalog No. NBP223475 Shop All Novus Products
Change view
Click to view available options
Buffer:
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buffer
NBP223475 Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223475 Supplier R&D Systems™ Supplier No. NBP223475
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. This product has been tested by SDS-PAGE and is ideal for blocking procedures such as Western Blotting, ELISA and immunochemistry to prevent nonspecific binding. Use in Immunohistochemistry-Paraffin reported in scientific literature (PMID: 26004262)

  • Lyophilized white powder or clear liquid.
  • Goat Serum blocking reagent/NGS was prepared from normal serum by multi-step process which includes delipidation, selective precipitation
  • Buffer: 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH: 7.2 and 0.01% (w/v) Gentamicin Sulfate
  • 0.01% Sodium Azide as preservative
  • Restore with deionized water (or equivalent)

Applications:

Western Blot, ELISA, Fluorophore-linked immunosorbent assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation

Recommended Storage

Store at 4°C for short term and -20°C for long term

Specifications

Buffer Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Purity or Quality Grade Multi-step Purification
Product Line Novus
Preservative 0.01% Sodium Azide, 0.01% Gentamicin Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.