Learn More
R&D Systems™ Nuclear Extraction Kit
SDP
Catalog No. NBP229447 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
NBP229447 1 Kit
1 options
Catalog No. NBP229447 Supplier R&D Systems™ Supplier No. NBP229447
Customers require a valid business account associated to their profile to access pricing and ordering options.
A Novus product, part of R&D Systems. Applications: Western Blot
Specifications
For Use With (Application) Western Blotting
Product Type Nuclear Extraction Kit
Content And Storage 10X Hypotonic Lysis Buffer 10mL, 1X Nuclear Extraction Buffer 10mL, 10% Detergent Solution 10mL, 10X PBS 2 x 50mL, 1 M DTT (for Nuclear Extraction from tissue) 100μL, 100X Protease Inhibitor Cocktail (PIC) 100μL, 100 mM PMSF 10mL. Storage is content dependent.
Product Line Novus
Quantity 1 Kit