Learn More
Novus Biologicals™ pCLNRX Retrovirus Expression Vector

Description
The pCLNRX expression vector is a part of the RetroMax expression system and has been designed to maximize recombinant-retrovirus titers in a simple, efficient, and flexible experimental system. All members of the RetroMax expression vector family (pCLXSN, pCLNCX, pCLNRX, and pCLNDX) have an extended packaging signal (omega+) and are derived from safety modified retrovirus vectors in which the gag open reading frame has been stopped by a point mutation, thereby minimizing the opportunity for replication competent retrovirus production by recombination with packaging genome.
- RetroMax system takes advantages of two properties of 293 cells (high level of transfectibility and strong E1A-mediated stimulation of CMV promoter controlled transcription)
- 293 cells of nonmurine origin avoid problem of selective packaging and transfer of VL30 genomes (present in all murine packaging cells)
- Vector supernatants are free of helper virus and are of sufficiently high titer within 2 days of transient transfection in 293 cells
- Buffer: 10μg in 20μL 1x TE (10mM Tris, pH 7.5, 1mM EDTA)
Recommended Storage
Store at -20°C. Avoid freeze-thaw cycles.
Specifications
Specifications
| Storage Buffer | 10µg in 20µL 1x TE (10mM Tris, pH 7.5, 1mM EDTA) |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 10 μg |
| Vector | pCLNRX Retrovirus Expression Vector |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.