Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ PDAP1 Protein
SDP

Catalog No. NBP1453105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1453105 0.5 mg
NBP14531001 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1453105 Supplier Novus Biologicals™ Supplier No. NBP1453100.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-181 of Human PDAP1 The Recombinant Human PDAP1 Protein is derived from E. coli. The Recombinant Human PDAP1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1.0mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 2mM DTT, 100mM NaCl, 0.1mM PMSF
Gene ID (Entrez) 11333
Molecular Weight (g/mol) 21.7kDa
Purification Method Protein
Quantity 0.5 mg
Source Human
Immunogen MPKGGRKGGH KGRARQYTSP EEIDAQLQAE KQKAREEEEQ KEGGDGAAGD PKKEKKSLDS DESEDEEDDY QQKRKGVEGL IDIENPNRVA QTTKKVTQLD LDGPKELSRR EREEIEKQKA KERYMKMHLA GKTEQAKADL ARLAIIRKQR EEAARKKEEE RKAKDDATLS GKRMQSLSLN KLEHHHHHH
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name PDAP1
Conjugate Unconjugated
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.