Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 494513 |
| Species | Human |
| Content And Storage | Store at 4C. After reconstitution store at -20°C. |
| Formulation | Lyophilized |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB110-75015. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | Autosomal recessive deafness type 59 protein, deafness, autosomal recessive 59, pejvakin, PJVK |
| Gene Symbol | DFNB59 |
| Product Type | Pejvakin |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
