Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 5730 |
| Species | Human |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB110-59911. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | Beta-trace protein, cerebrin-28, EC 5.3.99.2, Glutathione-independent PGD synthase, glutathione-independent PGD synthetase, lipocalin-type prostaglandin D synthase, Lipocalin-type prostaglandin-D synthase, L-PGDS, PDS, PGD2, PGD2 synthase, PGDS2, PGDSLPGDS, prostaglandin D synthase, prostaglandin D2 synthase (21 kD, brain), prostaglandin D2 synthase 21 kDa (brain), Prostaglandin-D2 synthase, prostaglandin-H2 D-isomerase |
| Gene Symbol | PTGDS |
| Product Type | PGD2 Synthase/PTGDS |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
