Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PI 3-Kinase p85 alpha N-terminal His (N-Term) Protein
SDP

Catalog No. NBP199881 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.005 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP199881 0.005 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP199881 Supplier Novus Biologicals™ Supplier No. NBP199881

Highly purified. Generating reliable and reproducible results.

Recombinant protein for Human PI 3-Kinase p85 alpha The Recombinant Human PI 3-Kinase p85 alpha Protein is derived from Sf 9 (baculovirus). The Recombinant Human PI 3-Kinase p85 alpha Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NM_181523
Formulation Recombinant protein in storage buffer (50mM sodium phosphate, pH 7.0, 300mM NaCl, 2mM DTT, 150mM imidazole, 0.1mM PMSF, 25 % glycerol.).mM
Gene ID (Entrez) 5295
Name PI 3-Kinase p85 alpha Protein
Purification Method SDS-PAGE
Quantity 0.005 mg
Source Baculovirus (Sf9 insect cells)
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Recombinant Recombinant
Research Category Cancer, Diabetes Research, Immune Dysfunction, Lipid and Metabolism, mTOR Pathway, Protein Kinase, Signal Transduction
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.