Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ PINK1 Protein
SDP

Catalog No. NBP17248110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17248110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17248110 Supplier Novus Biologicals™ Supplier No. NBP172481100UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged denatured recombinant protein corresponding to the amino acids 156-507 of Human PINK1 The Recombinant Human PINK1 Protein is derived from E. coli. The Recombinant Human PINK1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_115785
Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) 1M Urea, 5% Glycerol.
Gene ID (Entrez) 65018
Molecular Weight (g/mol) 37.9kDa
Purification Method SDS-PAGE
Quantity 100 μg
Source Recombinant human PINK protein was expressed in E. coli
Immunogen PINK1, 156-507 aa, Human, E. coli (NP_115785).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name PINK1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.