Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ QBRICK/FREM1 Antibody Blocking Peptide
SDP

Catalog No. NBP177180E Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177180E 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177180E Supplier Novus Biologicals™ Supplier No. NBP177180PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide containing 15 amino acids near the carboxy terminus of human FREM1. Source: Synthetic (Accession #: NP_659403) The QBRICK/FREM1 Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 158326
Species Human
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-77180. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias BNAR, C9orf143chromosome 9 open reading frame 154, C9orf145C9orf154extracellular matrix protein QBRICK, DKFZp686M16108, FLJ25461, FRAS1 related extracellular matrix 1, FRAS1-related extracellular matrix protein 1, Protein QBRICK, TILRR
Gene Symbol FREM1
Product Type QBRICK/FREM1
Quantity 0.05 mg
Regulatory Status RUO
Source Synthetic
Termination 15 amino acids near the carboxy terminus of human FRAM1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.