Learn More
Specifications
Specifications
| Target | QuikChIP |
| For Use With (Application) | Chromatin Immunoprecipitation |
| Product Type | Kit (ChIP) |
| Content And Storage | Wash Buffer A- 25mL, Wash Buffer B-25mL, Wash Buffer C- 25mL, Wash Buffer D- 50mL, SDS Lysis Buffer-15mL, 10X Glycine- 25mL, 10X PBS-55mL, ChIP Dilution Buffer- 50mL, Salmon Sperm DNA/ Protein A/G Agarose- 2 x 1.75mL, RNAse-10 mg/ml 60μL, 100X PMSF-0.75mL, 100X PIC-0.75mL, Proteinase K (10 mg/ml)-60μL, 20% SDS-625μL, 1M NaHCO3-1.25mL, 0.5M EDTA-250μL, 5M NaCl -500μL, 1M Tris-HCl- pH 6.5-500μL. Storage of components varies. See protocol for specific instructions. |
| Product Line | Novus |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
