Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ S100A9 Protein
SDP

Catalog No. NBP1445005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1445005 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP1445005 Supplier Novus Biologicals™ Supplier No. NBP1445000.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-114 of Human S100A9 The Recombinant Human S100A9 Protein is derived from E. coli. The Recombinant Human S100A9 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_002956
Concentration 1.0mg/mL
For Use With (Application) ELISA, In vivo Assay, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol 0.1M NaCl
Gene ID (Entrez) 6280
Molecular Weight (g/mol) 14.3kDa
Purification Method Protein
Quantity 0.5 mg
Source Recombinant S100A9 protein, fused to His-tag at C-terminus, expressed in E. coli
Immunogen S100A9, 1-114aa, Human, His tag, E. coli (NP_002956).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name S100A9
Conjugate Unconjugated
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.