Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ SNAP23 Protein
SDP

Catalog No. NBC11834702 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC11834702 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC11834702 Supplier Novus Biologicals™ Supplier No. NBC1183470.25MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-211 of Human SNAP23 The Recombinant Human SNAP23 Protein is derived from E. coli. The Recombinant Human SNAP23 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_003816.2
Concentration 1.0mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris pH 8.0
Gene ID (Entrez) 8773
Purification Method Protein
Quantity 0.25 mg
Source Recombinant SNAP23 protein expressed in E. coli
Immunogen SNAP23, 1-211aa, Human, Recombinant, E. coli (NP_003816.2).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name SNAP23
Conjugate Unconjugated
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.