Learn More
Novus Biologicals™ Preprotachykinin-1 Overexpression Lysate

Description
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_5474. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens Substance P, mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Target | Preprotachykinin-1 |
| Synonym | Hs0.2563, NK2, NKNA, NPK, PPT, protachykinin-1, TAC1, TAC2 |
| Protein State | Native |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.